6-Hydroxycoumarin ≥97%

Supplier: Matrix Scientific

Synonyms: 6-Hydroxy-2-benzopyrone, 6-Hydroxy-2H-chromen-2-one

085879-1G 085879-10G
102846-266EA 143.08 USD
102846-266 102846-268
6-Hydroxycoumarin ≥97%
6-Hydroxycoumarin

Hazard: Irritant

Formula: C₉H₆O₃
MW: 162.14 g/mol
MDL Number: MFCD00630040
CAS Number: 6093-68-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR