4-[(Allyloxy)methyl]-2,2-dimethyl-1,3-dioxolane ≥97%

Supplier: Matrix Scientific

085858-1G 085858-10G
102846-216EA 184.62 USD
102846-216 102846-218
4-[(Allyloxy)methyl]-2,2-dimethyl-1,3-dioxolane ≥97%
4-[(Allyloxy)methyl]-2,2-dimethyl-1,3-dioxolane

Hazard: Irritant

Formula: C₉H₁₆O₃
MDL Number: MFCD00099314
CAS Number: 4421-23-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR