(2Z)-Azepan-2-one oxime ≥97%
Supplier: Matrix Scientific
085845-1G
085845-10G
102846-184EA
200
USD
102846-184
102846-186
(2Z)-Azepan-2-one oxime ≥97%
(2Z)-Azepan-2-one oxime
Hazard: Irritant
Formula:
C₆H₁₂N₂O |
MDL Number:
MFCD00459957 CAS Number: 19214-08-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...