4-(Phenylsulfonyl)-2-azetidinone ≥97%

Supplier: Matrix Scientific

085842-1G 085842-10G 085842-50G
102846-168EA 101.54 USD
102846-168 102846-170 102846-172
4-(Phenylsulfonyl)-2-azetidinone ≥97%
4-(Phenylsulfonyl)-2-azetidinone

Hazard: Irritant

Formula: C₉H₉NO₃S
MW: 211.24 g/mol
MDL Number: MFCD00272351
CAS Number: 31899-01-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR