2-Thiohydantoin ≥97%

Supplier: Matrix Scientific

085841-1G 085841-10G 085841-50G
102846-162EA 56.92 USD
102846-162 102846-164 102846-166
2-Thiohydantoin ≥97%
2-Thiohydantoin

Hazard: Irritant

Formula: C₃H₄N₂OS
MDL Number: MFCD00005277
CAS Number: 503-87-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR