N-Cyclohexylguanidine hydrochloride ≥97%
Supplier: Matrix Scientific
085838-1G
085838-10G
102846-150EA
426.15
USD
102846-150
102846-152
N-Cyclohexylguanidine hydrochloride ≥97%
N-Cyclohexylguanidine hydrochloride
Hazard: Irritant
Formula:
C₇H₁₆ClN₃ |
MDL Number:
MFCD09800608 CAS Number: 2498-47-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...