(1S)-(+)-Camphorquinone ≥98%

Supplier: Matrix Scientific

Synonyms: (1S)-(+)-Camphorquin

085827-10G 085827-50G
102846-118EA 258.46 USD
102846-118 102846-120
(1S)-(+)-Camphorquinone ≥98%
(1S)-(+)-Camphorquinone

Hazard: Irritant

Formula: C₁₀H₁₄O₂
MW: 166.22 g/mol
MDL Number: MFCD00064159
CAS Number: 2767-84-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR