1-Oxaspiro[2.5]octane-2-carbonitrile ≥97%
Supplier: Matrix Scientific
085826-1G
085826-10G
102846-114EA
283.08
USD
102846-114
102846-116
1-Oxaspiro[2.5]octane-2-carbonitrile ≥97%
1-Oxaspiro[2.5]octane-2-carbonitrile
Hazard: Irritant
Formula:
C₈H₁₁NO |
MDL Number:
MFCD00101211 CAS Number: 36929-66-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...