4'-Methyl-1,1'-biphenyl-4-ol ≥97%

Supplier: Matrix Scientific

Synonyms: 4-Hydroxy-4'-methylbiphenyl

085823-1G 085823-10G
102846-104EA 298.46 USD
102846-104 102846-106
4'-Methyl-1,1'-biphenyl-4-ol ≥97%
4'-Methyl-1,1'-biphenyl-4-ol

Hazard: Irritant

Formula: C₁₃H₁₂O
MW: 184.24 g/mol
MDL Number: MFCD00218802
CAS Number: 26191-64-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR