1,4,7,10-Tetraazacyclododecane ≥98%
Supplier: Matrix Scientific
Synonyms:
Cyclen
085820-5G
085820-25G
102846-092EA
95.38
USD
102846-092
102846-094
1,4,7,10-Tetraazacyclododecane ≥98%
1,4,7,10-Tetraazacyclododecane
Hazard: Irritant
Formula:
C₈H₂₀N₄ MW: 172.27 g/mol |
MDL Number:
MFCD00066281 CAS Number: 294-90-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...