2,6-Dimethyl-4-pyrone ≥98%
Supplier: Matrix Scientific
Synonyms:
2,6-Dimethyl-4H-pyran-4-one, 2,6-Dimethyl-γ-pyrone
085817-1G
085817-10G
102846-082EA
72.31
USD
102846-082
102846-084
2,6-Dimethyl-4-pyrone ≥98%
2,6-Dimethyl-4-pyrone
Hazard: Irritant
Formula:
C₇H₈O₂ MW: 124.14 g/mol |
MDL Number:
MFCD00006579 CAS Number: 1004-36-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...