2-Cyclopenten-1-one ≥96%
Supplier: Matrix Scientific
Synonyms:
2-Cyclopentenone, Cyclopenten-2-one, Cyclopent-2-enone
085815-25G
085815-100G
102846-074EA
143.08
USD
102846-074
102846-076
2-Cyclopenten-1-one ≥96%
2-Cyclopenten-1-one
Hazard: Flammable
Formula:
C₅H₆O MW: 82.1 g/mol Boiling Pt: 150°C Density: 1.01 Flash Pt: 42°C |
MDL Number:
MFCD00001401 CAS Number: 930-30-3 UN: 1224 ADR: 3,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...