2-(Phenylthio)thiophene ≥97%

Supplier: Matrix Scientific

085812-1G 085812-10G 085812-50G
102846-058EA 123.08 USD
102846-058 102846-060 102846-062
2-(Phenylthio)thiophene ≥97%
2-(Phenylthio)thiophene

Hazard: Irritant

Formula: C₁₀H₈S₂
MW: 192.30 g/mol
MDL Number: MFCD00052141
CAS Number: 16718-12-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR