1-(2-Phenylethyl)homopiperazine ≥97%

Supplier: Matrix Scientific

085801-1G 085801-10G
102846-018EA 213.85 USD
102846-018 102846-020
1-(2-Phenylethyl)homopiperazine ≥97%
1-(2-Phenylethyl)homopiperazine

Hazard: Irritant

Formula: C₁₃H₂₀N₂
MDL Number: MFCD01320894
CAS Number: 40389-67-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR