Benzyl-3-oxocyclobutanecarboxylate ≥97%

Supplier: Matrix Scientific

Synonyms: 3-Oxo-cyclobutanecarboxylic acid benzyl ester, Benzyl 3-oxocyclobutanecarboxylate

085790-1G 085790-10G
102845-992EA 130.77 USD
102845-992 102845-994
Benzyl-3-oxocyclobutanecarboxylate ≥97%
Benzyl-3-oxocyclobutanecarboxylate

Hazard: Irritant

Formula: C₁₂H₁₂O₃
MW: 204.23 g/mol
MDL Number: MFCD11848894
CAS Number: 198995-91-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR