1-(4'-Pentyl-1,1'-biphenyl-4-yl)ethanone ≥97%

Supplier: Matrix Scientific

085781-1G 085781-10G
102845-972EA 184.62 USD
102845-972 102845-974
1-(4'-Pentyl-1,1'-biphenyl-4-yl)ethanone ≥97%
1-(4'-Pentyl-1,1'-biphenyl-4-yl)ethanone

Hazard: Irritant

Formula: C₁₉H₂₂O
MDL Number: MFCD00428365
CAS Number: 59662-38-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR