N-Vinyl-ε-caprolactam ≥97%
Supplier: Matrix Scientific
Synonyms:
1-Vinylazepan-2-one
085768-1G
085768-10G
085768-50G
102845-938EA
72.31
USD
102845-938
102845-940
102845-942
N-Vinyl-ε-caprolactam ≥97%
N-Vinyl-epsilon-caprolactam
Hazard: Irritant
Formula:
C₈H₁₃NO MW: 139.20 g/mol |
MDL Number:
MFCD00080693 CAS Number: 2235-00-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...