2-Acetylcyclohexanone ≥97%

Supplier: Matrix Scientific

Synonyms: 2-Acetyl-1-cyclohexanone

085757-1G 085757-10G
102845-912EA 33.85 USD
102845-912 102845-914
2-Acetylcyclohexanone ≥97%
2-Acetylcyclohexanone

Hazard: Irritant

Formula: C₈H₁₂O₂
MDL Number: MFCD00001633
CAS Number: 874-23-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR