Isocyanuricaciddiallylester ≥97%

Supplier: Matrix Scientific

085745-1G 085745-10G
102845-880EA 101.54 USD
102845-880 102845-882
Isocyanuricaciddiallylester ≥97%
Isocyanuricaciddiallylester

Hazard: Irritant

Formula: C₉H₁₁N₃O₃
MDL Number: MFCD00059763
CAS Number: 1081-69-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR