2-Mercapto-5-methyl-1,3,4-thiadiazole ≥97%

Supplier: Matrix Scientific

085726-1G 085726-10G 085726-50G
102845-832EA 364.62 USD
102845-832 102845-834 102845-836
2-Mercapto-5-methyl-1,3,4-thiadiazole ≥97%
2-Mercapto-5-methyl-1,3,4-thiadiazole

Hazard: Irritant

Formula: C₃H₄N₂S₂
MDL Number: MFCD00038349
CAS Number: 29490-19-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR