Pyrrolidine-2-thione ≥97%

Supplier: Matrix Scientific

085722-1G 085722-10G
102845-822EA 184.62 USD
102845-822 102845-824
Pyrrolidine-2-thione ≥97%
Pyrrolidine-2-thione

Hazard: Irritant

Formula: C₄H₇NS
MDL Number: MFCD00049351
CAS Number: 2295-35-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR