1-(3-Hydroxypropyl)-2-pyrrolidone ≥97%
Supplier: Matrix Scientific
Synonyms:
1-(3-Hydroxypropyl)pyrrolidin-2-one
085719-1G
085719-10G
102845-812EA
101.54
USD
102845-812
102845-814
1-(3-Hydroxypropyl)-2-pyrrolidone ≥97%
1-(3-Hydroxypropyl)-2-pyrrolidone
Hazard: Irritant
Formula:
C₇H₁₃NO₂ MW: 143.19 g/mol |
MDL Number:
MFCD00059014 CAS Number: 62012-15-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...