3-Methylpyrrolidin-2-one ≥97%

Supplier: Matrix Scientific

085718-1G 085718-10G
102845-808EA 496.92 USD
102845-808 102845-810
3-Methylpyrrolidin-2-one ≥97%
3-Methylpyrrolidin-2-one

Hazard: Irritant/Refrigerate

Formula: C₅H₉NO
Boiling Pt: 118-120°C/10 Torr
Melting Pt: 58…59 °C
MDL Number: MFCD00005271
CAS Number: 2555-05-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR