1-Ethyl-3-pyrrolidinol ≥97%

Supplier: Matrix Scientific

Synonyms: 1-Ethyl-3-hydroxypyrrolidine

085717-1G 085717-10G
102845-804EA 107.69 USD
102845-804 102845-806
1-Ethyl-3-pyrrolidinol ≥97%
1-Ethyl-3-pyrrolidinol

Hazard: Irritant

Formula: C₆H₁₃NO
MW: 115.18 g/mol
MDL Number: MFCD00003179
CAS Number: 30727-14-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR