3,5-Di(methylthio)isothiazole-4-carboxamide ≥97%

Supplier: Matrix Scientific

085713-1G 085713-10G
102845-790EA 241.54 USD
102845-790 102845-792
3,5-Di(methylthio)isothiazole-4-carboxamide ≥97%
3,5-Di(methylthio)isothiazole-4-carboxamide

Hazard: Irritant

Formula: C₆H₈N₂OS₃
MDL Number: MFCD00052563
CAS Number: 4886-14-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR