Piperonylonitrile ≥97%
Supplier: Matrix Scientific
Synonyms:
3,4-(Methylenedioxy)benzonitrile, 1,3-Benzodioxole-5-carbonitrile
085702-1G
085702-10G
085702-50G
102845-760EA
44.62
USD
102845-760
102845-762
102845-764
Piperonylonitrile ≥97%
Piperonylonitrile
Hazard: Irritant
Formula:
C₈H₅NO₂ MW: 147.13 g/mol Melting Pt: 94-97°C |
MDL Number:
MFCD00005820 CAS Number: 4421-09-4 UN: 3439 ADR: 6.1,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...