1-(2-Methylphenyl)piperazine dihydrochloride ≥97%
Supplier: Matrix Scientific
085697-1G
085697-10G
085697-50G
102845-746EA
101.54
USD
102845-746
102845-748
102845-750
1-(2-Methylphenyl)piperazine dihydrochloride ≥97%
1-(2-Methylphenyl)piperazine dihydrochloride
Hazard: Irritant
Formula:
C₁₁H₁₈Cl₂N₂ MW: 249.19 g/mol |
MDL Number:
MFCD00012759 CAS Number: 70849-60-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...