4-Piperazin-1-ylbutanenitrile ≥97%
Supplier: Matrix Scientific
085696-1G
085696-10G
102845-742EA
112.31
USD
102845-742
102845-744
4-Piperazin-1-ylbutanenitrile ≥97%
4-Piperazin-1-ylbutanenitrile
Hazard: Irritant
Formula:
C₈H₁₅N₃ |
MDL Number:
MFCD03093804 CAS Number: 5623-94-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...