1-(2,3-Xylyl)piperazin ≥98%

Supplier: Matrix Scientific

Synonyms: 1-(2,3-Xylyl)piperazine, 1-(2,3-Dimethylphenyl)piperazine

085693-1G 085693-10G
102845-732EA 72.31 USD
102845-732 102845-734
1-(2,3-Xylyl)piperazin ≥98%
1-(2,3-Xylyl)piperazin

Hazard: Irritant

Formula: C₁₂H₁₈N₂
MW: 190.29 g/mol
MDL Number: MFCD00040730
CAS Number: 1013-22-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR