1-(3-Phenypropyl)piperazine ≥97%
Supplier: Matrix Scientific
085692-1G
085692-10G
102845-728EA
86.15
USD
102845-728
102845-730
1-(3-Phenypropyl)piperazine ≥97%
1-(3-Phenypropyl)piperazine
Hazard: Irritant
Formula:
C₁₃H₂₀N₂ |
MDL Number:
MFCD01075188 CAS Number: 55455-92-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...