1-(1-Phenylethyl)piperazine ≥97%

Supplier: Matrix Scientific

085690-1G 085690-10G
102845-720EA 53.85 USD
102845-720 102845-722
1-(1-Phenylethyl)piperazine ≥97%
1-(1-Phenylethyl)piperazine

Hazard: Irritant

Formula: C₁₂H₁₈N₂
MDL Number: MFCD00040738
CAS Number: 69628-75-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR