1-(6-Methylpyridin-2-yl)piperazine ≥97%

Supplier: Matrix Scientific

085689-1G 085689-10G
102845-716EA 130.77 USD
102845-716 102845-718
1-(6-Methylpyridin-2-yl)piperazine ≥97%
1-(6-Methylpyridin-2-yl)piperazine

Hazard: Irritant

Formula: C₁₀H₁₅N₃
MW: 177.25 g/mol
MDL Number: MFCD01320891
CAS Number: 55745-89-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR