2-Piperazin-1-ylphenol dihydrobromide ≥97%
Supplier: Matrix Scientific
Synonyms:
1-(2-Hydroxyphenyl)piperazine dihydrobromide
085688-1G
085688-10G
085688-50G
102845-710EA
38.46
USD
102845-710
102845-712
102845-714
2-Piperazin-1-ylphenol dihydrobromide ≥97%
2-Piperazin-1-ylphenol dihydrobromide
Hazard: Irritant
Formula:
C₁₀H₁₆Br₂N₂O |
MDL Number:
MFCD00040814 CAS Number: 58260-69-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...