cis-2,6-Dimethylpiperazine ≥97%
Supplier: Matrix Scientific
085687-1G
085687-10G
085687-50G
102845-704EA
72.31
USD
102845-704
102845-706
102845-708
cis-2,6-Dimethylpiperazine ≥97%
cis-2,6-Dimethylpiperazine
Hazard: Irritant
Formula:
C₆H₁₄N₂ MW: 114.19 g/mol |
MDL Number:
MFCD07772435 CAS Number: 21655-48-1 UN: 1325 ADR: 4.1,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...