1-(4-tert-Butylbenzyl)piperazine ≥97%
Supplier: Matrix Scientific
085685-1G
085685-10G
102845-698EA
101.54
USD
102845-698
102845-700
1-(4-tert-Butylbenzyl)piperazine ≥97%
1-(4-tert-Butylbenzyl)piperazine
Hazard: Irritant
Formula:
C₁₅H₂₄N₂ |
MDL Number:
MFCD00082594 CAS Number: 956-61-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...