2-(Piperazin-1-yl)pyrazine ≥97%

Supplier: Matrix Scientific

Synonyms: 1-(2-Pyrazinyl)piperazine

085682-1G 085682-10G
102845-688EA 101.54 USD
102845-688 102845-690
2-(Piperazin-1-yl)pyrazine ≥97%
2-(Piperazin-1-yl)pyrazine

Hazard: Irritant

Formula: C₈H₁₂N₄
MW: 164.212 g/mol
MDL Number: MFCD00040766
CAS Number: 34803-68-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR