1-Propylpiperazine dihydrobromide ≥97%

Supplier: Matrix Scientific

085681-1G 085681-10G
102845-684EA 72.31 USD
102845-684 102845-686
1-Propylpiperazine dihydrobromide ≥97%
1-Propylpiperazine dihydrobromide

Hazard: Irritant

Formula: C₇H₁₆N₂
MW: 290.04 g/mol
MDL Number: MFCD00051992
CAS Number: 64262-23-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR