1-(5-Chloro-2-methylphenyl)piperazine ≥97%
Supplier: Matrix Scientific
085678-1G
085678-10G
102845-672EA
176.92
USD
102845-672
102845-674
1-(5-Chloro-2-methylphenyl)piperazine ≥97%
1-(5-Chloro-2-methylphenyl)piperazine
Hazard: Irritant
Formula:
C₁₁H₁₅ClN₂ |
MDL Number:
MFCD00087128 CAS Number: 76835-20-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...