1-(Cyclohexylmethyl)piperazine ≥97%
Supplier: Matrix Scientific
085675-1G
085675-10G
102845-662EA
127.69
USD
102845-662
102845-664
1-(Cyclohexylmethyl)piperazine ≥97%
1-(Cyclohexylmethyl)piperazine
Hazard: Irritant
Formula:
C₁₁H₂₂N₂ MW: 182.311 g/mol |
MDL Number:
MFCD01075237 CAS Number: 57184-23-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...