1-(2-Pyrimidyl)piperazine ≥97%

Supplier: Matrix Scientific

Synonyms: 2-(1-Piperazinyl)pyrimidine, 2-(Piperazin-1-yl)pyrimidine

085672-1G 085672-10G
102845-654EA 56.92 USD
102845-654 102845-656
1-(2-Pyrimidyl)piperazine ≥97%
1-(2-Pyrimidyl)piperazine

Hazard: Irritant

Formula: C₈H₁₂N₄
MW: 164.21 g/mol
MDL Number: MFCD00040742
CAS Number: 20980-22-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR