1-Benzyl-4-cyano-4-hydroxypiperidine ≥97%

Supplier: Matrix Scientific

085649-1G 085649-10G
102845-592EA 21.54 USD
102845-592 102845-594
1-Benzyl-4-cyano-4-hydroxypiperidine ≥97%
1-Benzyl-4-cyano-4-hydroxypiperidine

Hazard: Irritant

Formula: C₁₃H₁₆N₂O
MW: 216.29 g/mol
Melting Pt: 95…102 °C
MDL Number: MFCD00023752
CAS Number: 6094-60-6
UN: 3439
ADR: 6.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR