2-Phenyl-2-(2-piperidyl)acetamide hydrate ≥97%

Supplier: Matrix Scientific

085647-1G 085647-10G
102845-586EA 127.69 USD
102845-586 102845-588
2-Phenyl-2-(2-piperidyl)acetamide hydrate ≥97%
2-Phenyl-2-(2-piperidyl)acetamide hydrate

Hazard: Irritant

Formula: C₁₃H₁₈N₂O
MDL Number: MFCD06656234
CAS Number: 19395-39-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR