4-Hydroxy-4-phenylpiperidine ≥97%
Supplier: Matrix Scientific
Synonyms:
4-Phenyl-4-piperidinol
085645-1G
085645-10G
102845-580EA
127.69
USD
102845-580
102845-582
4-Hydroxy-4-phenylpiperidine ≥97%
4-Hydroxy-4-phenylpiperidine
Hazard: Irritant
Formula:
C₁₁H₁₅NO MW: 177.25 g/mol |
MDL Number:
MFCD00006000 CAS Number: 40807-61-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...