5-Chloro-1-(4-piperidinyl)-2-benzimidazolidinone ≥97%
Supplier: Matrix Scientific
Synonyms:
5-Chloro-1-(piperidin-4-yl)-1H-benzo[d]imidazol-2(3H)-one, 5-Chloro-1-(4-piperidinyl)-2-benzimidazolinone
085634-1G
085634-10G
102845-548EA
76.92
USD
102845-548
102845-550
5-Chloro-1-(4-piperidinyl)-2-benzimidazolidinone ≥97%
5-Chloro-1-(4-piperidinyl)-2-benzimidazolidinone
Hazard: Irritant
Formula:
C₁₂H₁₄ClN₃O MW: 251.72 g/mol |
MDL Number:
MFCD02093793 CAS Number: 53786-28-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...