4-Benzoylpiperidine hydrochloride ≥97%
Supplier: Matrix Scientific
Synonyms:
Phenyl(piperidin-4-yl)methanone hydrochloride
085627-1G
085627-10G
102845-532EA
200
USD
102845-532
102845-534
4-Benzoylpiperidine hydrochloride ≥97%
4-Benzoylpiperidine hydrochloride
Hazard: Irritant
Formula:
C₁₂H₁₆ClNO MW: 225.7200 g/mol |
MDL Number:
MFCD00066982 CAS Number: 25519-80-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...