1-Boc-2-cyanopiperidine ≥97%
Supplier: Matrix Scientific
Synonyms:
tert-Butyl 2-cyanopiperidine-1-carboxylate
085609-1G
085609-10G
102845-492EA
200
USD
102845-492
102845-494
1-Boc-2-cyanopiperidine ≥97%
1-Boc-2-cyanopiperidine
Hazard: Irritant
Formula:
C₁₁H₁₈N₂O₂ MW: 210.27 g/mol |
MDL Number:
MFCD01861221 CAS Number: 153749-89-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...