2-(Piperidinomethyl)acrylonitrile ≥95%
Supplier: Matrix Scientific
085602-1G
085602-10G
102845-474EA
110.77
USD
102845-474
102845-476
2-(Piperidinomethyl)acrylonitrile ≥95%
2-(Piperidinomethyl)acrylonitrile
Hazard: Corrosive/Stored under Argon/Moisture Sensitive
Formula:
C₉H₁₄N₂ |
MDL Number:
MFCD00052023 CAS Number: 27315-95-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...