tert-Butyl-4-pyrrolidin-1-ylpiperidine-1-carboxylate ≥97%

Supplier: Matrix Scientific

085599-1G 085599-10G
102845-466EA 467.69 USD
102845-466 102845-468
tert-Butyl-4-pyrrolidin-1-ylpiperidine-1-carboxylate ≥97%
tert-Butyl-4-pyrrolidin-1-ylpiperidine-1-carboxylate

Hazard: Irritant

Formula: C₁₄H₂₆N₂O₂
MW: 254.38 g/mol
MDL Number: MFCD08056347
CAS Number: 902837-26-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR