2-(1-Piperazinylcarbonyl)-1,4-benzodioxane hydrochloride ≥97%

Supplier: Matrix Scientific

Synonyms: 1-[(1,4-Benzodioxan-2-yl)carbonyl]piperazine Hydrochloride, 2-(1-Piperazinylcarbonyl)-1,4-benzodioxane Hydrochloride

085587-1G 085587-10G
102845-434EA 101.54 USD
102845-434 102845-436
2-(1-Piperazinylcarbonyl)-1,4-benzodioxane hydrochloride ≥97%
2-(1-Piperazinylcarbonyl)-1,4-benzodioxane hydrochloride

Hazard: Irritant

Formula: C₁₃H₁₆N₂O₃
MW: 284.74 g/mol
MDL Number: MFCD00729051
CAS Number: 70918-74-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR