Decahydroisoquinoline ≥97%
Supplier: Matrix Scientific
085569-1G
085569-10G
102845-392EA
69.23
USD
102845-392
102845-394
Decahydroisoquinoline ≥97%
Decahydroisoquinoline
Hazard: Irritant
Formula:
C₉H₁₇N |
MDL Number:
MFCD00012096 CAS Number: 6329-61-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...